missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93867-0.02ml
This item is not returnable.
View return policy
Description
RNPC3 Polyclonal antibody specifically detects RNPC3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RNPC3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ20008, FLJ25070, KIAA183, KIAA1839, RBM40U11/U12 snRNP 65 kDa protein, RNA recognition protein, RNA-binding motif protein 40, RNA-binding protein 40, RNA-binding region (RNP1, RRM) containing 3, RNA-binding region-containing protein 3, RNP, U11/U12 small nuclear ribonucleoprotein 65 kDa protein, U11/U12 snRNP 65K, U11/U12-65K | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 258-517 of human RNPC3 (NP_060089.1). NKLMELANLQPKRPKTIKQRHVRKKRKIKDMLNTPLCPSHSSLHPVLLPSDVFDQPQPVGNKRIEFHISTDMPAAFKKDLEKEQNCEEKNHDLPATEVDASNIGFGKIFPKPNLDITEEIKEDSDEMPSECISRRELEKGRISREEMETLSVFRSYEPGEPNCRIYVKNLAKHVQEKDLKYIFGRYVDFSSETQRIMFDIRLMKEGRMKGQAFIGLPNEKAAAKALKEANGYVLFGKPMVVQFARSARPKQDPKEGKRKC | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55599 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction