missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNPC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107249
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 92%; Dog: 92%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 92%; Rat: 85%; Yeast: 77%.
Participates in pre-mRNA U12-dependent splicing, performed by the minor spliceosome which removes U12-type introns. U12-type introns comprises less than 1% of all non-coding sequences. Binds to the 3'-stem-loop of m(7)G-capped U12 snRNA.
Specifications
| RNPC3 | |
| Polyclonal | |
| Unconjugated | |
| RNPC3 | |
| 2810441O16Rik; AI447568; C030014B17Rik; KIAA1839; Rbm40; RNA binding region (RNP1, RRM) containing 3; RNA recognition protein; RNA-binding motif protein 40; RNA-binding protein 40; RNA-binding region (RNP1, RRM) containing 3; RNA-binding region-containing protein 3; RNP; Rnpc3; SNRNP65; U11/U12 small nuclear ribonucleoprotein 65 kDa protein; U11/U12 snRNP 65 kDa protein; U11/U12 snRNP 65K; U11/U12-65K | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q96LT9 | |
| RNPC3 | |
| Synthetic peptide directed towards the middle region of human RNPC3 (aa 215-265). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction