missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88168
This item is not returnable.
View return policy
Description
RNPS1 Polyclonal specifically detects RNPS1 in Human samples. It is validated for Western Blot.
Specifications
| RNPS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| E5.1, LDC2, MGC117332, RNA binding protein S1, serine-rich domain, RNA-binding protein S1, serine-rich domain, RNA-binding protein with serine-rich domain 1, SR protein, SR-related protein LDC2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human RNPS1 | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10921 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction