Learn More
Tocris Bioscience™ Ro 48-8071 fumarate
2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis
Brand: Tocris Bioscience™ 5389/50
Description
Ro 48-8071 fumarate is a 2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).Specifications
| 99% | |
| 189197-69-1 | |
| 50 mg |
| (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone | |
| 4°C |