missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO1 Antibody (770502) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB71181X
This item is not returnable.
View return policy
Description
ROBO1 Monoclonal antibody specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| ROBO1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 770502 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO1, Met1-Ala858, Accession # Q9Y6N7 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 6091 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction