Learn More
Novus Biologicals™ ROBO1 Antibody (Asclepius Technology patent anti-Robo1 CAR) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28772-100ug
 View more versions of this product
Product Code. 30047750
  • € 338.00 / 100µg
Description
ROBO1 Monoclonal antibody specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ROBO1
Monoclonal
Unconjugated
Q9Y6N7
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Asclepius Technology patent anti-Robo1 CAR
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
ROBO1
100 μg
Angiogenesis, Cancer, Extracellular Matrix
6091