missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROBO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575814
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specialized cells at the midline, which separates the left and right halves of the CNS, have a number of roles in directing growth cone behavior. In the vertebrate spinal cord, the insect ventral nerve cord and in C. elegans, midline cells produce guidance cues such as nectins and slit, which act as attractants and repellents, respectively. These cells may act as gatekeepers to prevent axons from crossing the midline and to induce a switch in growth cone responsiveness to guidance cues beyond the gateway. One such gatekeeper, Robo, is an axon guidance receptor that defines a novel subfamily of Ig superfamily proteins that are conserved from fruit flies to mammals. Robo acts as a receptor for the repellent Slit and functions in a cell-autonomous fashion. Non-crossing axons express high levels of Robo, whereas crossing axons express low levels of Robo before reaching the midline and high levels after they cross. Robo1 and Robo2 are two human homologs of the Drosophila protein Roundabout. Robo1 is also homologous to the C. elegans gene sax3, whereas Robo2 is homologous to the zebrafish gene astray.
Specifications
| ROBO1 | |
| Polyclonal | |
| Unconjugated | |
| ROBO1 | |
| AW494633; AW742721; deleted in U twenty twenty; DUTT1; FLJ21882; Gm310; HGNC:10249; H-Robo-1; MGC131599; MGC133277; ROBO1; roundabout 1; roundabout guidance receptor 1; roundabout homolog 1; roundabout homolog 1 (Drosophila); roundabout, axon guidance receptor, homolog 1; roundabout, axon guidance receptor, homolog 1 (Drosophila); Roundabout1 protein; SAX3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19876, 58946, 6091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O55005, O89026, Q9Y6N7 | |
| ROBO1 | |
| Synthetic peptide corresponding to residues in Human ROBO1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction