missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO2 Antibody (356001), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3147PEATT594
This item is not returnable.
View return policy
Description
ROBO2 Monoclonal antibody specifically detects ROBO2 in Human samples. It is validated for Flow Cytometry
Specifications
| ROBO2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6092 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 356001 | |
| Flow Cytometry | |
| Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila) | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO2 isoform 1 Ser22-Pro859 Accession # Q9HCK4.2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction