Learn More
Invitrogen™ ROBO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587613
Description
Immunogen sequence: TALLKCKATG DPLPVISWLK EGFTFPGRDP RATIQEQGTL QIKNLRISDT GTYTCVATSS SGETSWSAVL DVTESGATIS KNYDLSDLPG PPSKPQVTDV TKNSVTLSWQ PGTPGTLPAS AYIIEAFSQS VSNSWQTVAN HVKTTLYTVR GLRPNTIYLF MVRAINPQGL SDPSPMSDPV RTQDISPPAQ GVDHRQVQKE LGDVLVRLHN P; Positive Samples: SH-SY5Y, SKOV3, A-549, Mouse brain, Mouse lung, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.
ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| ROBO2 | |
| Polyclonal | |
| Unconjugated | |
| ROBO2 | |
| 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 268902, 6092, 84409 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TPD3, Q9HCK4 | |
| ROBO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-660 of human ROBO2 (NP_0011224011). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.