missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROBO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144712
This item is not returnable.
View return policy
Description
Immunogen sequence: HLDELTRAYQ FDIAKQTWHI QSNNQPPQPP VPPLGYVSGA LISDLETDVA DDDADDEEEA LEIPRPLRAL DQTPGSSMDN LDSSVTGKAF TSSQRPRPTS PFSTDSNTSA ALSQSQRPRP TKKHKGGRMD QQPALPHRRE GM Antigen sequence identity: Mouse 92%; Rat 92%.
ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| ROBO2 | |
| Polyclonal | |
| Unconjugated | |
| ROBO2 | |
| 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCK4 | |
| ROBO2 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1168-1309. Recombinant protein control fragment (Product #RP-109728). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction