missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ROBO2 Pre-design Chimera RNAi

Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variants mRNA. This product targets for all of 5 transcript variants (NM_001128929,NM_001290039,NM_001290040,NM_001290065,NM_002942)

Brand:  Abnova™ H00006092-R04.10nmol

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16028958

  • € 189.00 / 10nmol


Estimated Shipment
03-11-2020
Add to basket

Description

Description

  • Gene name: ROBO2
  • Gene Alias: KIAA1568,SAX3
  • Entrez GeneID: 6092
  • strong>Gene Description: Roundabout, axon guidance receptor, homolog 2 (Drosophila)
Specifications

Specifications

RNAi Knockdown
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variant 4, mRNA.
ROBO2 Pre-design Chimera RNAi
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity.
Human
DEPC water
10 nmol
Human
Videos
SDS
Documents

For research use only