missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO4 Antibody (036), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90787MFV610
This item is not returnable.
View return policy
Description
ROBO4 Monoclonal antibody specifically detects ROBO4 in Mouse samples. It is validated for ELISA
Specifications
| ROBO4 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 036 | |
| ELISA | |
| ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ROBO4 (Accession#: NP_083059.2; Met1-Glu480). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix, Neuroscience | |
| 54538 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction