missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO4 Antibody (274914) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB50041JF585
This item is not returnable.
View return policy
Description
ROBO4 Monoclonal antibody specifically detects ROBO4 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| ROBO4 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
| Y3 rat myeloid cell line transfected with mouse ROBO4, Leu28-Ser1012, Accession # Q8C310 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 274914 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix, Neuroscience | |
| 54538 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction