missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO4 Monoclonal Antibody (OTI4E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811323
This item is not returnable.
View return policy
Description
Roundabout 4 is receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. This protein may mediate the inhibition of primary endothelial cell migration by Slit proteins.Specifications
| ROBO4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WZ75 | |
| ROBO4 | |
| Human recombit protein fragment corresponding to amino acids 32-331 of human ROBO4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4E2 | |
| Unconjugated | |
| ROBO4 | |
| ECSM4, MRB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54538 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction