missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Human Naïve Pan T Cell Isolation Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Human Naïve Pan T Cell Isolation Kit is designed to isolate naïve pan T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
Specifications
| Cell Type | T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction