missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RoboSep™ Mouse Hematopoietic Progenitor Cell Isolation Kit

Product Code. 17117981
Encompass
Change view
Click to view available options
:
1 x 109 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17117981 1 x 109 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17117981 Supplier STEMCELL Technologies Supplier No. 19856RF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips

  • The EasySep™ Mouse Hematopoietic Cell Isolation Kit is designed to isolate stem and progenitor cells from single-cell suspensions of bone marrow or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-hematopoietic stem cells and non-progenitor cells (CD5, CD11b, CD19, CD45R/B220, Ly6G/C(Gr-1), TER119, 7-4) and streptavidin-coated magnetic particles (RapidSpheres™ )
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This product replaces the EasySep™ Mouse Hematopoietic Progenitor Cell Enrichment Kit (Catalog #19756) for faster cell isolations

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.