Learn More
Testo™ Robust Food Probe
Click to view available options
Diameter (Metric) Probe:
4 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Type | Probe, Robust Food |
| Certifications/Compliance | 60751, EN 13485 and HACCP-compliant |
| Response Time | 10 sec. |
| Diameter (Metric) Probe | 4 mm |