Learn More
ROCK1 Recombinant Rabbit Monoclonal Antibody (23GB2065), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen MA552474
 View more versions of this product
Product Code. 30088674
  • € 468.00 / 100µL
Description
ROCK1 Recombinant Monoclonal antibody specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ROCK1
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P70335, Q13464, Q63644
Rock1
A synthesized peptide derived from human ROCK1 (1100-1200AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2065
Unconjugated
Rock1
1110055K06Rik; Ac2-154; Liver regeneration-related protein LRRG199; MGC131603; MGC43611; p150 RhoA-binding kinase ROK beta; p160 ROCK-1; p160ROCK; p160-ROCK; PRO0435; Renal carcinoma antigen NY-REN-35; Rho associated coiled-coil containing protein kinase 1; Rho kinase; Rho-associated coiled-coil containing protein kinase 1; Rho-associated coiled-coil forming kinase 1; Rho-associated kinase beta; rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK1; ROCK-I
Rabbit
Affinity Chromatography
RUO
19877, 6093, 81762
-20°C