missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROCK1/ROCK2 Recombinant Rabbit Monoclonal Antibody (HL1632)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550324
This item is not returnable.
View return policy
Description
ROCK1/ROCK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ROCK1 and ROCK2 are genes that encode for Rho-associated coiled-coil containing protein kinases, also known as ROCKs. These kinases are involved in a variety of cellular processes such as cell migration, adhesion, proliferation, and apoptosis. They are activated by the small GTPase RhoA and phosphorylate downstream targets such as myosin light chain, LIM kinase, and myosin phosphatase. Dysregulation of ROCK1 and ROCK2 has been linked to various diseases including cancer, cardiovascular disease, and neurological disorders.
Specifications
| ROCK1/ROCK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O75116, Q13464 | |
| ROCK1, ROCK2 | |
| Recombinant fragment of human ROCK1 . | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL1632 | |
| Unconjugated | |
| ROCK2 | |
| ROCK-1/ROCK-2; ROCK-I/ROCK-II | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6093, 9475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction