missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-593B
This item is not returnable.
View return policy
Description
ROCK2 Polyclonal antibody specifically detects ROCK2 in Human samples. It is validated for Western Blot
Specifications
| ROCK2 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2 | |
| The immunogen recognized by this antibody maps to a region between residues 1320 and the C-terminus (residues 1388) of human ROCK2 using the number given in entry NP_004841.1 (GeneID 9475) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase, Wnt Signaling Pathway | |
| 9475 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction