Learn More
SP Genevac™ Rocket Synergy 2 Outer Lid Seal
Product Code. 31723017
Change view
Click to view available options
Material:
EPDM
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
31723017 EPDM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31723017 Supplier SP Genevac™ Supplier No. 045818/S
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement seal for the for the outer Lid of the Genevac Rocket Synergy 2 Benchtop Evaporator
  • Replacement Seal
  • Compatible with Rocket Synergy 2 Rocket Synergy Rocket 4D Synergy and Pre Synergy Rocket Evaporators
Specifications
Material EPDM
Type Seal