missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROGDI Polyclonal Antibody
GREENER_CHOICE

Product Code. 16646664
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16646664 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16646664 Supplier Invitrogen™ Supplier No. PA5100179

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ROGDI.

This gene encodes a protein of unknown function. Loss-of-function mutation in this gene cause Kohlschutter-Tonz syndrome. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROGDI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ROGDI
Gene Accession No. Q3TDK6, Q9GZN7
Gene Alias 0610011C19Rik; AU020118; C76152; KTZS; leucine zipper domain protein; leucine-zipper-containing LZF; Lzf; protein rogdi homolog; ROGDI; rogdi atypical leucine zipper; rogdi homolog; rogdi homolog (Drosophila)
Gene Symbols ROGDI
Host Species Rabbit
Immunogen A synthesized peptide derived from human ROGDI(Accession Q9GZN7), corresponding to amino acid residues L196-K246.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66049, 79641
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.