missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Rolipram

Product Code. p-7217659
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 17160212

Brand: STEMCELL Technologies 73382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cAMP pathway activator; Inhibits type 4 cyclic nucleotide phosphodiesterases (PDE4)

  • Rolipram is a cell-permeable, selective inhibitor of Type 4 cyclic nucleotide phosphodiesterases (PDE4), which mediate cyclic AMP (cAMP) degradation
  • Rolipram preferably inhibits PDE4 isoform A (IC50 = 3 nM) over other isoforms such as B and D (IC50 = 130 and 240 nM respectively; MacKenzie & Houslay)
  • It inhibits interferon (IFN)-γ stimulated phosphorylation of p38 mitogen-activated protein (MAP) kinase through PDE4B and/or PDE4D isoform inhibition (MacKenzie & Houslay)

Specifications

CAS 61413-54-5
Synonym SB 95952; ZK 62711
Quantity 5 mg
Molecular Formula C16H21NO3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.