missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROMO1 Monoclonal Antibody (OTI5G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505611
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a mitochondrial membrane protein that is responsible for increasing the level of reactive oxygen species (ROS) in cells. The protein also has antimicrobial activity against a variety of bacteria by inducing bacterial membrane breakage.Specifications
| ROMO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P60602 | |
| ROMO1 | |
| Full length human recombit protein of human ROMO1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5G8 | |
| Unconjugated | |
| ROMO1 | |
| C20orf52, MTGM, MTGMP, bA353C18.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 140823 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction