missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROPN1L Polyclonal Antibody
GREENER_CHOICE

Product Code. 15797591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15797591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15797591 Supplier Invitrogen™ Supplier No. PA559746

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PDTMFCAQQI HIPPELPDIL KQFTKAAIRT QPADVLRWSA GYFSALSRGD PLPVKDRMEM PTATQKTDTG LTQGLL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Principal Names: ROPN1L; Ropporin-1 like; ASP; AKAP-associated sperm protein; Official Gene Symbol: ROPN1L Gene ID: 83853 (human) Gene Map Locus: 5p15.2 (human) AKAPs act as scaffolding proteinscoordinating phosphatase/kinase signaling at the cytoskeleton. They interact with PKAs and anchor them to specific sub-cellular compartments and participate in multiple cellular processes including growth, metabolism and differentiation. ASP, a 26 kDa protein, is a novel AKAP-interacting protein. It interacts with the amphipathic helix of AKAP3 and facilitates AKAP3-PKA interactions. It consists of a conserved N-terminal AKAP-binding domain and a central cysteine-rich Fz domain. It is specifically expressed in the sperms and might play a significant role in sperm motility and also facilitate cytoskeletal reorganization during spermiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROPN1L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ropn1l
Gene Accession No. Q96C74
Gene Alias Akapasp; AKAP-associated sperm protein; A-kinase anchoring protein-associated sperm protein; ASP; AV047578; radial spoke head 11 homolog; rhophilin associated tail protein 1 like; rhophilin associated tail protein 1-like; ROPN1L; ROPN1-like protein; ropporin 1-like; ropporin-1-like protein; RSPH11
Gene Symbols Ropn1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ROPN1L. Recombinant protein control fragment (Product #RP-102916).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.