missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Roquin Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81891AF488
This item is not returnable.
View return policy
Description
Roquin Polyclonal antibody specifically detects Roquin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Roquin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| KIAA2025RP5-1198E17.5, RING finger and C3H zinc finger protein 1, ring finger and CCCH-type domains 1, RING finger protein 198, RNF198ring finger and CCCH-type zinc finger domains 1, roquin | |
| Rabbit polyclonal Roquin antibody was raised against an 18 amino acid peptide near the carboxy terminus of human Roquin. The immunogen is located within amino acids 1040 - 1090 of Roquin. Amino Acid Squence: KLNTSQQAENGQPEPQNK | |
| 0.1 mL | |
| Immunology | |
| 149041 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction