missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR beta Antibody (OTI1G1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73930IR
This item is not returnable.
View return policy
Description
ROR beta Monoclonal specifically detects ROR beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ROR beta | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RORB | |
| Recombinant protein fragment corresponding to amino acids 1-260 of human ROR beta (NP_008845) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1G1 | |
| Western Blot, Immunohistochemistry | |
| NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 6096.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction