missing translation for 'onlineSavingsMsg'
Learn More

ROR beta Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97604PCP

Product Code. 18656512

  • ONLINE EXCLUSIVE PRICE
    € 430.00
    € 406.35 / 0.10mL
    Save € 23.65
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ROR beta Polyclonal antibody specifically detects ROR beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ROR beta
Polyclonal
Immunohistochemistry-Paraffin
NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA
Produced in rabbits immunized with E. coli-derived Human ROR beta fragment.
0.1 mL
Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction
6096
Store at 4C in the dark.
IgG
Immunohistochemistry (Paraffin)
PerCP
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.