missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR gamma/RORC/NR1F3 Antibody (4G419), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24449JF646
This item is not returnable.
View return policy
Description
ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ROR gamma/RORC/NR1F3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RORC | |
| A portion of amino acids 1-50 of human ROR gamma was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| 4G419 | |
| Western Blot, Flow Cytometry | |
| MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 6097.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction