missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR gamma/RORC/NR1F3 Antibody (RORC/8017R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20738AF405
This item is not returnable.
View return policy
Description
ROR gamma/RORC/NR1F3 Monoclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ROR gamma/RORC/NR1F3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
| Recombinant full-length human ROR gamma/RORC/NR1F3 protein | |
| 0.1 mL | |
| GPCR | |
| 6097 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| RORC/8017R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction