missing translation for 'onlineSavingsMsg'
Learn More

ROR gamma (t) Monoclonal Antibody (B2D), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6981-82

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15257957

  • € 518.00 / 100µg
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The B2D monoclonal antibody reacts with the mouse RORgamma(t) protein. RORgamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes RORalpha and RORbeta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. RORgamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, RORgammat, which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ "e;double-positive"e; thymoyctes, Th17 cells of the periphery and lymphoid tissue inducer (Lti) cells of lymphoid organs. The RORgammat isoform differs from RORgamma by three unique amino acids at the amino terminus. Therefore, the B2D antibody will react with both the RORgamma and RORgammat isoforms. Applications Reported: This B2D antibody has been reported for use in immunoblotting (WB). Applications Tested: This B2D antibody has been tested by western blot analysis of mouse thymus nuclear lysates. This B2D antibody can be used at a starting concentration of 2 μg/mL, however the optimal concentration should be determined for individual experimental conditions. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

ROR gamma, a NR1 Thyroid Hormone-Like receptor, has been shown to affect thymopoiesis, bone metabolism, T-cell apoptosis, and lymphoid organogenesis. ROR gamma has been shown to promote thymocyte survival by activating the expression of the antiapoptotic protein Bcl-x(L). ROR gamma is also required for the development of lymph nodes and Peyer's patches. It has been shown that ROR gamma(t), a thymus-specific isoform of ROR gamma from mouse, inhibits Fas ligand expression and cytokine secretion in immature thymocytes. ROR gamma binds as a monomer to response elements composed of a single core motif, GGTCA, preceded by a 6 bp AT-rich sequence. ROR gamma expression has been documented in mouse thymus, adipose, bone, skeletal muscle, liver, and kidney, and in human skeletal muscle. ESTs have been isolated from human tissue libraries, including cancerous head/neck, germ cell, and prostate, and normal colon, head/neck, lung, liver/spleen, and pancreas. ROR gammat t (RORgt) is an isoform of RORC.
Specifications

Specifications

ROR gamma (t)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P51450
Rorc
Affinity chromatography
RUO
19885
4° C
Liquid
Western Blot
B2D
Unconjugated
Rorc
EGK_01272; IMD42; MGC129539; NR1F3; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; Rorc; Rorg; RORgamma; RORgt; RP11-98D18.11-001; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR
Rat
100 μg
Primary
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.