missing translation for 'onlineSavingsMsg'
Learn More

ROR gamma t/RORC2/NR1F3 Antibody (1181A), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals FAB61093H

Product Code. 18629793

  • € 639.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ROR gamma t/RORC2/NR1F3 Monoclonal antibody specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ROR gamma t/RORC2/NR1F3
Monoclonal
HRP
PBS
Rabbit
0.1 mL
Primary
Human
IgG
Flow Cytometry
1181A
Intracellular Staining by Flow Cytometry
IMD42;NR1F3;Nuclear receptor ROR-gamma;RORG;RZRG;RZR-GAMMA;TOR
E.coli derived recombinant human ROR gamma T/RORC2/NR1F3, Met1-Gln100, Accession # P51449
RUO
6097
Store at 4C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.