missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR1 Antibody (zilovertamab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28157JF669
This item is not returnable.
View return policy
Description
ROR1 Monoclonal antibody specifically detects ROR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ROR1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
| ROR1 | |
| 0.1 mL | |
| Protein Kinase | |
| 4919 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| zilovertamab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction