missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (ROR2/1911), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08532PCP
This item is not returnable.
View return policy
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ROR2 | |
| Monoclonal | |
| PerCP | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Human cultured keratinocytes (Uniprot: Q01974) | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 4920 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ROR2/1911 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction