missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Mouse, Clone: Ror2, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565550
This item is not returnable.
View return policy
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human, Mouse , Rat samples. It is validated for Western blot .
Spezifikation
| ROR2 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Ror2 | |
| Unconjugated | |
| Neurotrophic tyrosine kinase, receptor-related 2; NTRKR2, BDB, BDB1 | |
| Mouse ROR2 Peptide | |
| RUO | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Reseach Use Only.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur