missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110312
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: CMRIGIPAER LGRYHQCYNG SGMDYRGTAS TTKSGHQCQP WALQHPHSHH LSSTDFPELG GGHAYCRNPG GQMEGPWCFT QNKNVRMELC DVPSCSPRDS S.
ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors. It is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development. This Type I membrane protein is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Defects in ROR2 are a cause of brachydactyly type B1 (BDB1). BDB1 is an autosomal dominant skeletal disorder characterized by hypoplasia/aplasia of distal phalanges and nails. In BDB1 the middle phalanges are short but in addition the terminal phalanges are rudimentary or absent. Both fingers and toes are affected. The thumbs and big toes are usually deformed. Defects in ROR2 are a cause of recessive Robinow syndrome (RRS). RRS is an autosomal disorder characterized by skeletal dysplasia with generalized limb bone shortening, segmental defects of the spine, brachydactyly and a dysmorphic facial appearance. The protein contains 1 frizzled (FZ) domain, 1 immunoglobulin-like C2-type domain, and 1 kringle domain.
Tekniske data
| ROR2 | |
| Polyclonal | |
| Unconjugated | |
| ROR2 | |
| BDB; BDB1; mRor2; neurotrophic tyrosine kinase receptor-related 2; neurotrophic tyrosine kinase, receptor related 2; neurotrophic tyrosine kinase, receptor-related 2; NTRKR2; receptor tyrosine kinase like orphan receptor 2; receptor tyrosine kinase-like orphan receptor 2; ROR2; Tyrosine-protein kinase transmembrane receptor ROR2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 306782 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.85 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 0 | |
| ROR2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-400 of human ROR2 (NP_004551.2). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion