missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORA Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803201
This item is not returnable.
View return policy
Description
RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).Specifications
| RORA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35398 | |
| RORA | |
| Human recombit protein fragment corresponding to amino acids 98-345 of human RORA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| RORA | |
| NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6095 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction