Learn More
RORC Mouse anti-Human, Clone: OTI1G3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809592
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RORC (RAR related orphan receptor C) is a DNA-binding transcription factor and is a member of the NR1 subfamily of nuclear hormone receptors. The specific functions of RORC are not known; however, studies of a similar gene in mice have shown that this gene may be essential for lymphoid organogenesis and may play an important regulatory role in thymopoiesis. In addition, studies in mice suggest that the protein encoded by the RORC gene may inhibit the expression of Fas ligand and IL2. Diseases associated with RORC include Immunodeficiency 42 and Anus Cancer.Specifications
| RORC | |
| Monoclonal | |
| Unconjugated | |
| RORC | |
| IMD42; NR1F3; RORG; RZR-GAMMA; RZRG; TOR; RORgt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6097 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1G3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P51449 | |
| RORC | |
| Human recombit protein fragment corresponding to amino acids 1-266 of human RORC produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.