missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS Antibody (5-4E), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50180AF647
This item is not returnable.
View return policy
Description
ROS Monoclonal specifically detects ROS in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ROS | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ROS1 | |
| Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography. | |
| 0.1 mL | |
| Protein Kinase | |
| 6098 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 5-4E | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1 receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction