missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS Antibody (OTI1F3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73932DL594
This item is not returnable.
View return policy
Description
ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| ROS | |
| Monoclonal | |
| DyLight 594 | |
| proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
| Recombinant protein fragment corresponding to amino acids 2126-2347 of human ROS (NP_002935) produced in E.coli. | |
| 0.1 mL | |
| Protein Kinase | |
| 6098 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1F3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction