Learn More
STEMCELL Technologies RosetteSep™ HLA B Cell Enrichment Cocktail
Description
- The RosetteSep™ HLA B Cell Enrichment Cocktail is designed to isolate B cells from whole blood by negative selection.
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and red blood cells (RBCs).
- When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs.
- The purified B cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
- The desired cells are immediately ready for serology or flow cytometry crossmatch assays, or other downstream applications.
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Cell Isolation; In Vitro Diagnostic |
| Quantity | 1000 mL blood |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.