missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ HLA Total Lymphocyte Enrichment Cocktail

Product Code. 17187691
Encompass
Click to view available options
Quantity:
1000 mL whole blood
250 mL whole blood
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17187691

Brand: STEMCELL Technologies 15283HLA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity isolation of untouched lymphocytes for HLA analysis.

  • The RosetteSep™ HLA Total Lymphocyte Enrichment Cocktail is designed to enrich lymphocytes from whole blood by negative selection.
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD16, CD36, CD66b and glycophorin A on red blood cells (RBCs).
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs.
  • The purified lymphocytes are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
  • The desired cells are immediately ready for flow cytometry crossmatch assays or other downstream applications.

Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.

Specifications

Cell Type Lymphocytes
For Use With (Application) Cell Isolation; In Vitro Diagnostic
Quantity 1000 mL whole blood
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.