Learn More
Rosiglitazone Maleate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Rosiglitazone Maleate
Specifications
Specifications
| Chemical Name or Material | Rosiglitazone Maleate |
| CAS | 155141-29-0 |
| Analyte or Component Names | Rosiglitazone Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N3 O3 S . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2,4-Thiazolidinedione, 5-[[4-[2-(methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-, (2Z)-2-butenedioate (1:1), 2,4-Thiazolidinedione, 5-[[4-[2-(methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-, (Z)-2-butenedioate (1:1), 5-[[4-[2-(N-Methyl-N-2-pyridylamino)ethoxy]phenyl]methyl]thiazolidine-2,4-dione maleate, Avandia, BRL 49653C, Rosiglitazone maleate, BRL-49653C, SB214521, SB236833, SB237216, BRL 49653C |
| InChI Formula | InChI=1S/C18H19N3O3S.C4H4O4/c1-21(16-4-2-3-9-19-16)10-11-24-14-7-5-13(6-8-14)12-15-17(22)20-18(23)25-15;5-3(6)1-2-4(7)8/h2-9,15H,10-12H2,1H3,(H,20,22,23);1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN(CCOc1ccc(CC2SC(=O)NC2=O)cc1)c3ccccn3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione |
| Show More |