Learn More
Quickfit™ Plain Side-Arms
Product Code. 12957306
Change view
Click to view available options
Diameter (Metric):
8 mm
10 mm
13 mm
Unit Size:
Pack of 10
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
12957306 10 mm Pack of 5
12485266 8 mm Pack of 5
15577910 13 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12957306 Supplier Quickfit Supplier No. GP6RA/7
Only null left
Add to basket
Add to basket
Design gives excellent flow control and allows use with fluids and gases having temperatures between -20°C and 200°C.
  • Rotary design for fine single drop control invaluable on burettes
  • Borosilicate glass with PTFE key for excellent chemical resistance
  • No lubrication required - no seizure and no contamination
  • Temperature range -20°C to 200°C
  • Vacuum capability to 10-4Torr
  • Increased barrel diameter for excellent drainage
  • Locking ring to prevent accidental removal
  • Ergonomic key for good grip and easy operation
TRUSTED_SUSTAINABILITY
Specifications
Type Plain Side-arm
Diameter (Metric) 10 mm
Material Borosilicate Glass, PTFE (Key)
For Use With (Equipment) GP Rotaflo™ stopcock