Learn More
Vacuubrand™ Rotary Vane Pump RZ 9
GREENER_CHOICE
Product Code. 17856238
Click to view available options
Plug Type:
CEE 7/7
Unit Size:
Each
Product Code. 17856238
Brand: Vacuubrand™ 20798140
Only null left
Add to basket
Add to basket
Rotary vane pumps offer high performance in a compact design. Even with the gas ballast open, they enable an ideal ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to its optimized lubrication circuit and large usable oil volume.
  • Very high flow rates, even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast
  • Ideal ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume allows for long intervals between oil changes
  • Ease of maintenance due to telescopic design
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 2 x 10-3 mbar
Dimensions (L x W x H) 503 x 156 x 240 mm
Plug Type CEE 7/7
Flow Rate 8.9 m³/hr.
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP40
Applications Drying Chambers, Lyophilization, Fine Vacuum Distillation
Maximum Pressure 1.1 bar
Voltage 230 V