Learn More
Corning™ Rotor Adapter for LSE™ Mini Microcentrifuge 6770-RT
Shop All Corning Products
Click to view available options
Capacity:
0.2 mL
0.25 mL
0.5 mL
Unit Size:
Pack of 8
Description
Corning™ Rotor Adapter comes in three sizes and is for use with Corning Mini Microcentrifuge 6770
Specifications
Specifications
| Type | Rotor Adapter |
| Capacity | 0.2 mL |
| For Use With (Equipment) | Corning™ Mini Microcentrifuge 6770 |
| No. per Pack | 8/Pk. |