Learn More
Hettich Lab Technology™ Swing-out Rotor for Manual Centrifuge
Click to view available options
Capacity:
4 x 15 mL
Unit Size:
Each
Description
- 90° angle
- Max. RCF of 1,298
- Max. RPM of 3,000/min.
- 4 places for 15 mL conical tubes
Specifications
Specifications
| Type | Swing-out Rotor |
| Capacity | 4 x 15 mL |
| Max. RCF | 1,298 x g |
| Max. Speed | 3,000 rpm |
| For Use With (Equipment) | Hettich Hand Centrifuge with Head |
| Angle | 90° |
| No. of Places | 4 |