missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotundine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Rotundine |
| CAS | 483-14-7 |
| Molecular Formula | C21 H25 N O4 |
| Quantity | 100 mg |
| Synonym | 13aα-Berbine','2,3,9,10-tetramethoxy- (8CI), 6H-Dibenzo[a,g]quinolizine','5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy-','(S)-, Cyclanoline','O,O-dimethyl-N-demethyl- (6CI), Gindarine (7CI), (13aS)-5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, (-)-2,3,9,10-Tetramethoxyberbine, (-)-Corydalis B, (-)-Rotundine, (-)-S-Tetrahydropalmatine, (-)-Tetrahydropalmatine, (13AS)-2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline, (S)-Tetrahydropalmatine, Caseanine, Hyndarine, N-Demethyl-O,O'-dimethylcyclanoline, N-Demethyl-O,O'-dimethylsteponine, Palmatine','tetrahydro-','(-)-, Rotundine, Rotundine (Stephania)','(-)-, l-Tetrahydropalmatine |
| InChI Formula | InChI=1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3/t17-/m0/s1 |
| SMILES | COc1cc2CCN3Cc4c(C[C@H]3c2cc1OC)ccc(OC)c4OC |
| IUPAC Name | (13aS)-2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinoline |
| Molecular Weight (g/mol) | 355.43 |
| Formula Weight | 355.1784 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction