Learn More
Lenz Laborglasintrumente Round Bottom Flasks
Product Code. 10274101
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1 L
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Diameter (Metric) Outer Neck:
34 mm
50 mm
51 mm
65 mm
76 mm
89 mm
Unit Size:
Each
Pack of 10
Product Code. 10274101
Brand: Lenz Laborglasintrumente 07035073
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Diameter (Metric) Outer Neck 50 mm
Capacity (Metric) 2 L
Material Duran Glass
Height (Metric) 240 mm