missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Round Bottom Flasks

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07035084

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11973621

  • € 180.20 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications

Specifications

89 mm
6 L
Duran Glass
330 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers